Assay Details
| Gene Symbol: Cr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005265.1 | NP_001005265.1 | 7 - 8 | 1312 | 87 | |
| NM_001005330.1 | NP_001005330.1 | 1312 | 87 | |||
| NM_019301.2 | NP_062174.1 | 7 - 8 | 1312 | 87 | ||
| GenBank mRNA | BC061736.1 | - | 7 - 8 | 1081 | 5599318 | 87 |
| D42114.1 | - | 7 - 8 | 1121 | 87 | ||
| D42115.1 | - | 7 - 8 | 1057 | 87 | ||
| D42116.1 | - | 4 - 5 | 665 | 87 | ||
| FQ220559.1 | - | 3 - 4 | 331 | 87 | ||
| L36532.1 | - | 7 - 8 | 1080 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cr1l
Genomic Map