Assay Details
| Gene Symbol: Prl8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022846.2 | NP_074037.1 | 1 - 2 | 38 | 118 | |
| GenBank mRNA | FQ229765.1 | - | 1 - 2 | 95 | 118 | |
| L06441.1 | - | 1 - 2 | 40 | 118 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the L06441.1,NM_022846.2 transcripts.
Set Membership: |
|

Prl8a2
Genomic Map