Assay Details
| Gene Symbol: Rab3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031091.2 | NP_112353.1 | 2 - 3 | 340 | 61 | |
| XM_006238531.2 | XP_006238593.1 | 2 - 3 | 386 | 61 | ||
| GenBank mRNA | BC060562.1 | - | 2 - 3 | 258 | 6918181 | 61 |
| U68184.1 | - | 1 - 2 | 46 | 61 | ||
| Y14019.1 | - | 1 - 2 | 228 | 61 | ||
More Information
Set Membership: |
|

Rab3b
Genomic Map