Assay Details
| Gene Symbol: Kcns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053954.1 | NP_446406.1 | 1 - 2 | 78 | 63 | |
| GenBank mRNA | Y17606.1 | - | 1 - 2 | 78 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_053954.1, Y17606.1 transcripts.
Set Membership: |
|

Kcns1
Genomic Map