Assay Details
| Gene Symbol: Capn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133309.2 | NP_579843.2 | 4 - 5 | 611 | 63 | |
| XM_006250363.2 | XP_006250425.2 | 4 - 5 | 63 | |||
| GenBank mRNA | D14478.1 | - | 5 - 6 | 692 | 63 | |
| D14479.1 | - | 4 - 5 | 611 | 63 | ||
| D14480.1 | - | 4 - 5 | 597 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Capn8
Genomic Map