Assay Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270871.1 | NP_001257800.1 | 17 - 18 | 1677 | 79 | |
| NM_001270872.1 | NP_001257801.1 | 16 - 17 | 1641 | 79 | ||
| NM_181366.2 | NP_852031.1 | 18 - 19 | 1725 | 79 | ||
| XM_006256916.2 | XP_006256978.1 | 18 - 19 | 1765 | 79 | ||
| XM_008773154.1 | XP_008771376.1 | 17 - 18 | 2105 | 79 | ||
| XM_008773155.1 | XP_008771377.1 | 17 - 18 | 1715 | 79 | ||
| XM_008773156.1 | XP_008771378.1 | 16 - 17 | 2069 | 79 | ||
| XM_008773157.1 | XP_008771379.1 | 16 - 17 | 2063 | 79 | ||
| XM_008773158.1 | XP_008771380.1 | 15 - 16 | 2027 | 79 | ||
| XM_008773159.1 | XP_008771381.1 | 14 - 15 | 1618 | 79 | ||
| XM_008773160.1 | XP_008771382.1 | 17 - 18 | 2106 | 79 | ||
| GenBank mRNA | AF538953.1 | - | 18 - 19 | 1735 | 79 | |
| AF538958.1 | - | 17 - 18 | 1687 | 79 | ||
| AF538959.1 | - | 16 - 17 | 1651 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr64
Genomic Map