Assay Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270871.1 | NP_001257800.1 | 22 - 23 | 2322 | 79 | |
| NM_001270872.1 | NP_001257801.1 | 21 - 22 | 2286 | 79 | ||
| NM_181366.2 | NP_852031.1 | 23 - 24 | 2370 | 79 | ||
| XM_006256916.2 | XP_006256978.1 | 23 - 24 | 2410 | 79 | ||
| XM_008773154.1 | XP_008771376.1 | 22 - 23 | 2750 | 79 | ||
| XM_008773155.1 | XP_008771377.1 | 22 - 23 | 2360 | 79 | ||
| XM_008773156.1 | XP_008771378.1 | 21 - 22 | 2714 | 79 | ||
| XM_008773157.1 | XP_008771379.1 | 21 - 22 | 2708 | 79 | ||
| XM_008773158.1 | XP_008771380.1 | 20 - 21 | 2672 | 79 | ||
| XM_008773159.1 | XP_008771381.1 | 19 - 20 | 2263 | 79 | ||
| XM_008773160.1 | XP_008771382.1 | 22 - 23 | 2751 | 79 | ||
| GenBank mRNA | AF538953.1 | - | 23 - 24 | 2380 | 79 | |
| AF538958.1 | - | 22 - 23 | 2332 | 79 | ||
| AF538959.1 | - | 21 - 22 | 2296 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr64
Genomic Map