Assay Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270871.1 | NP_001257800.1 | 6 - 7 | 330 | 109 | |
| NM_001270872.1 | NP_001257801.1 | 5 - 6 | 294 | 109 | ||
| NM_181366.2 | NP_852031.1 | 7 - 8 | 378 | 109 | ||
| XM_006256916.2 | XP_006256978.1 | 7 - 8 | 418 | 109 | ||
| XM_008773154.1 | XP_008771376.1 | 6 - 7 | 758 | 109 | ||
| XM_008773155.1 | XP_008771377.1 | 6 - 7 | 368 | 109 | ||
| XM_008773156.1 | XP_008771378.1 | 5 - 6 | 722 | 109 | ||
| XM_008773159.1 | XP_008771381.1 | 3 - 4 | 271 | 109 | ||
| XM_008773160.1 | XP_008771382.1 | 6 - 7 | 759 | 109 | ||
| GenBank mRNA | AF538953.1 | - | 7 - 8 | 388 | 109 | |
| AF538958.1 | - | 6 - 7 | 340 | 109 | ||
| AF538959.1 | - | 5 - 6 | 304 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr64
Genomic Map