Assay Details
| Gene Symbol: Qrfp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198200.1 | NP_937843.1 | 1 | 257 | 58 | |
| XM_008761671.1 | XP_008759893.1 | 1 | 270 | 58 | ||
| GenBank mRNA | AB109627.1 | - | 1 | 257 | 58 | |
| AY438327.1 | - | 1 | 263 | 58 | ||
| Gene Symbol: LOC100909783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749440.2 | XP_003749488.1 | 1 | 270 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qrfp
Genomic Map