Assay Details
| Gene Symbol: Tox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199392.2 | NP_955424.2 | 3 - 4 | 451 | 82 | |
| XM_006235578.2 | XP_006235640.1 | 3 - 4 | 485 | 82 | ||
| GenBank mRNA | AB096685.1 | - | 3 - 4 | 452 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tox2
Genomic Map