Assay Details
| Gene Symbol: Ns5atp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201418.1 | NP_958821.1 | 3 - 4 | 289 | 136 | |
| GenBank mRNA | AY496944.1 | - | 3 - 4 | 289 | 136 | |
| FQ220183.1 | - | 3 - 4 | 366 | 136 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ns5atp9
Genomic Map