Assay Details
| Gene Symbol: Ldhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017266.2 | NP_058962.2 | 6 - 7 | 811 | 73 | |
| XM_006229233.2 | XP_006229295.1 | 6 - 7 | 882 | 73 | ||
| XM_008759344.1 | XP_008757566.1 | 6 - 7 | 758 | 73 | ||
| XM_008759345.1 | XP_008757567.1 | 5 - 6 | 584 | 73 | ||
| GenBank mRNA | BC078862.1 | - | 6 - 7 | 811 | 7131609 | 73 |
| JQ173138.1 | - | 4 - 5 | 534 | 73 | ||
| JQ173139.1 | - | 3 - 4 | 100 | 73 | ||
| JQ173140.1 | - | 6 - 7 | 799 | 73 | ||
| JQ409364.1 | - | 4 - 5 | 534 | 73 | ||
| JQ409365.1 | - | 3 - 4 | 100 | 73 | ||
| JQ409366.1 | - | 6 - 7 | 799 | 73 | ||
| JQ409368.1 | - | 3 - 4 | 344 | 73 | ||
| U07177.1 | - | 5 - 6 | 712 | 73 | ||
More Information
Set Membership: |
|

Ldhc
Genomic Map