Assay Details
| Gene Symbol: Tpsab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019322.2 | NP_062195.2 | 1 - 2 | 273 | 77 | |
| GenBank mRNA | BC061983.1 | - | 1 - 2 | 255 | 5598863 | 77 |
| U67910.1 | - | 1 - 2 | 272 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpsab1
Genomic Map