Assay Details
| Gene Symbol: Cmklr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022218.2 | NP_071554.2 | 2 | 1142 | 104 | |
| XM_006249505.2 | XP_006249567.1 | 3 | 1515 | 104 | ||
| XM_006249506.2 | XP_006249568.1 | 3 | 1194 | 104 | ||
| XM_006249507.2 | XP_006249569.1 | 3 | 1841 | 104 | ||
| XM_006249508.2 | XP_006249570.1 | 3 | 1458 | 104 | ||
| XM_006249509.2 | XP_006249571.1 | 3 | 1358 | 104 | ||
| GenBank mRNA | AJ002745.1 | - | 1 | 1049 | 104 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cmklr1
Genomic Map