Assay Details
| Gene Symbol: Rem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022685.2 | NP_073176.2 | 2 - 3 | 534 | 65 | |
| GenBank mRNA | AF084464.1 | - | 2 - 3 | 408 | 65 | |
| AY916790.1 | - | 2 - 3 | 474 | 65 | ||
| BC127495.1 | - | 2 - 3 | 519 | 8371030 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rem2
Genomic Map