Assay Details
| Gene Symbol: Arl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031711.1 | NP_113899.1 | 1 - 2 | 101 | 98 | |
| XM_006230899.1 | XP_006230961.1 | 1 - 2 | 150 | 98 | ||
| GenBank mRNA | FQ213929.1 | - | 1 - 2 | 109 | 98 | |
| Y12708.1 | - | 1 - 2 | 101 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arl2
Genomic Map