Assay Details
| Gene Symbol: Trpv5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053787.2 | NP_446239.2 | 12 - 13 | 1661 | 103 | |
| XM_006236376.2 | XP_006236438.1 | 10 - 11 | 1237 | 103 | ||
| GenBank mRNA | AB032019.1 | - | 12 - 13 | 1661 | 103 | |
| AF209196.1 | - | 1612 | 103 | |||
| AY762624.1 | - | 11 - 12 | 1349 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpv5
Genomic Map