Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134387.1 | NP_599214.1 | 1 - 2 | 66 | 61 | |
| XM_008768465.1 | XP_008766687.1 | 1 - 2 | 112 | 61 | ||
| XM_008768466.1 | XP_008766688.1 | 1 - 2 | 112 | 61 | ||
| XM_008768467.1 | XP_008766689.1 | 1 - 2 | 112 | 61 | ||
| GenBank mRNA | AB061719.1 | - | 1 - 2 | 66 | 61 | |
| FQ233401.1 | - | 1 - 2 | 65 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dcxr
Genomic Map