Assay Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138885.1 | NP_620240.1 | 6 - 7 | 813 | 58 | |
| XM_006248379.2 | XP_006248441.1 | 6 - 7 | 2683 | 58 | ||
| XM_006248380.2 | XP_006248442.1 | 5 - 6 | 2531 | 58 | ||
| XM_008768698.1 | XP_008766920.1 | 5 - 6 | 1876 | 58 | ||
| XM_008768699.1 | XP_008766921.1 | 5 - 6 | 1876 | 58 | ||
| XM_008768700.1 | XP_008766922.1 | 5 - 6 | 1876 | 58 | ||
| XM_008768701.1 | XP_008766923.1 | 5 - 6 | 1876 | 58 | ||
| XM_008768702.1 | XP_008766924.1 | 4 - 5 | 525 | 58 | ||
| XM_008768703.1 | XP_008766925.1 | 5 - 6 | 1877 | 58 | ||
| GenBank mRNA | D25543.1 | - | 6 - 7 | 813 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golgb1
Genomic Map