Assay Details
| Gene Symbol: Lgals3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139096.1 | NP_620796.1 | 3 - 4 | 402 | 88 | |
| GenBank mRNA | AF065438.1 | - | 3 - 4 | 402 | 88 | |
| BC081724.1 | - | 3 - 4 | 375 | 7123606 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgals3bp
Genomic Map