Assay Details
| Gene Symbol: G6pc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176077.3 | NP_788266.2 | 5 - 6 | 847 | 83 | |
| GenBank mRNA | AY186240.1 | - | 6 - 7 | 677 | 83 | |
| BC078689.1 | - | 5 - 6 | 847 | 7134166 | 83 | |
| FQ231610.1 | - | 5 - 6 | 743 | 83 | ||
More Information
Set Membership: |
|

G6pc3
Genomic Map