Assay Details
| Gene Symbol: Stx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031125.1 | NP_112387.1 | 3 - 4 | 268 | 65 | |
| XM_006230345.2 | XP_006230407.1 | 3 - 4 | 775 | 65 | ||
| XM_006230346.2 | XP_006230408.1 | 3 - 4 | 800 | 65 | ||
| XM_006230348.2 | XP_006230410.1 | 3 - 4 | 765 | 65 | ||
| GenBank mRNA | L20821.1 | - | 3 - 4 | 268 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stx4
Genomic Map