Assay Details
| Gene Symbol: Acsm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144748.1 | NP_653349.1 | 1 - 2 | 98 | 106 | |
| XM_008759678.1 | XP_008757900.1 | 1 - 2 | 120 | 106 | ||
| XM_008759679.1 | XP_008757901.1 | 1 - 2 | 121 | 106 | ||
| GenBank mRNA | AF062389.1 | - | 1 - 2 | 98 | 106 | |
| BC078721.1 | - | 1 - 2 | 75 | 7110246 | 106 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF062389.1,BC078721.1,NM_144748.1,XM_008759678.1,XM_008759679.1 transcripts.
Set Membership: |
|

Acsm2a
Genomic Map