Assay Details
| Gene Symbol: Bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182674.3 | NP_872615.3 | 3 - 4 | 634 | 115 | |
| GenBank mRNA | AY257675.4 | - | 3 - 4 | 617 | 115 | |
| BC079030.1 | - | 3 - 4 | 623 | 7114086 | 115 | |
More Information
Set Membership: |
|

Bysl
Genomic Map