Assay Details
| Gene Symbol: Mthfs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009349.1 | NP_001009349.1 | 1 - 2 | 123 | 107 | |
| GenBank mRNA | BC087144.1 | - | 1 - 2 | 123 | 7128624 | 107 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
metabolic process
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of apoptotic process
tetrahydrofolate metabolic process
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
molecular_function
ATP binding
5-formyltetrahydrofolate cyclo-ligase activity
protein homodimerization activity
metal ion binding
protein heterodimerization activity
biological_process
metabolic process
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of apoptotic process
tetrahydrofolate metabolic process
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of intrinsic apoptotic signaling pathway
molecular_function
ATP binding
5-formyltetrahydrofolate cyclo-ligase activity
protein homodimerization activity
metal ion binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P001219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55810 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001219 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P001219 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P001219 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P935273 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P18403 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45827 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85267 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72674 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19896 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10912118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912116 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10912115 | - | - |
| Rat Genome 230 2.0 Array | 1392719_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392719_at | - | - |
| Rat Genome 230 2.0 Array | 1381768_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381768_at | - | - |
| Rat Genome 230 2.0 Array | 1384370_at | - | - |
| Rat Genome 230 2.0 Array | 1384370_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381768_at | - | - |
| Affymetrix HT Rat Focus Array | 1392719_at | - | - |
| Affymetrix HT Rat Focus Array | 1381768_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392719_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384370_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1384370_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800009230.rn.1 | TC0800001265.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800009226.rn.1 | TC0800001265.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800009228.rn.1 | TC0800001265.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800009229.rn.1 | TC0800001265.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800009232.rn.1 | TC0800001265.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800009231.rn.1 | TC0800001265.rn.1 | RTA TC |
| Rat Expression Set 230 | 1381768_at | - | - |
| Rat Expression Set 230 | 1392719_at | - | - |
| Rat Expression Set 230 | 1381768_PM_at | - | - |
| Rat Expression Set 230 | 1384370_at | - | - |
| Rat Expression Set 230 | 1392719_PM_at | - | - |
| Rat Expression Set 230 | 1384370_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10912115 | - | - |
| Rat Genome U34 Set | rc_AI178809_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358377 | - | - |
| RatRef-12 Expression BeadChip | ILMN_51113 | - | - |

Mthfs
Genomic Map