Assay Details
| Gene Symbol: Pxylp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007710.1 | NP_001007711.1 | 2 - 3 | 172 | 62 | |
| XM_006243591.2 | XP_006243653.1 | 2 - 3 | 609 | 62 | ||
| XM_006243592.2 | XP_006243654.1 | 2 - 3 | 1712 | 62 | ||
| GenBank mRNA | BC081981.1 | - | 2 - 3 | 172 | 7128571 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Pxylp1
Genomic Map