Assay Details
| Gene Symbol: Csrnp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271225.1 | NP_001258154.1 | 1 - 2 | 73 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271225.1 transcripts.
Set Membership: |
|

Csrnp3
Genomic Map