Assay Details
| Gene Symbol: Ckap2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001057508.5 | XP_001057508.3 | 2 - 3 | 176 | 72 | |
| XM_006234995.2 | XP_006235057.1 | 2 - 3 | 176 | 72 | ||
More Information
Set Membership: |
|

Ckap2l
Genomic Map