Assay Details
| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234810.2 | XP_006234872.1 | 7 - 8 | 629 | 57 | |
| XM_008762122.1 | XP_008760344.1 | 7 - 8 | 629 | 57 | ||
More Information
Set Membership: |
|

Pla2g4f
Genomic Map