Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107761.1 | NP_001101231.1 | 6 - 7 | 1243 | 67 | |
| XM_006234766.2 | XP_006234828.1 | 6 - 7 | 1004 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Exd1
Genomic Map