Assay Details
| Gene Symbol: Chac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173437.1 | NP_001166908.1 | 1 - 2 | 421 | 78 | |
| GenBank mRNA | EF688602.1 | - | 1 - 2 | 421 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chac1
Genomic Map