Assay Details
| Gene Symbol: Plxna4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762859.1 | XP_008761081.1 | 17 - 18 | 2363 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plxna4a| Gene Symbol: Plxna4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762859.1 | XP_008761081.1 | 17 - 18 | 2363 | 85 | |
Set Membership: |
|