Assay Details
| Gene Symbol: Henmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037655.1 | NP_001032744.2 | 88 | 102 | ||
| GenBank mRNA | BC107926.1 | - | 88 | 7132526 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107926.1 transcripts.
Set Membership: |
|

Henmt1
Genomic Map