Assay Details
| Gene Symbol: Aknad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044303.2 | NP_001037768.2 | 5 - 6 | 1422 | 82 | |
| NR_073134.1 | 5 - 6 | 1422 | 82 | |||
| XM_008761422.1 | XP_008759644.1 | 5 - 6 | 2603 | 82 | ||
| GenBank mRNA | BC079051.1 | - | 5 - 6 | 1422 | 7115381 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Aknad1
Genomic Map