Assay Details
| Gene Symbol: F3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013057.2 | NP_037189.2 | 1 - 2 | 182 | 110 | |
| GenBank mRNA | BC081846.1 | - | 1 - 2 | 182 | 7096602 | 110 |
More Information
Updated Mapping Information
Set Membership: |
|

F3
Genomic Map