Assay Details
| Gene Symbol: LOC100909548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003752100.1 | XP_003752148.1 | 4 - 5 | 617 | 60 | |
| XM_006257201.2 | XP_006257263.1 | 4 - 5 | 1663 | 60 | ||
| XM_006257202.1 | XP_006257264.1 | 4 - 5 | 836 | 60 | ||
| Gene Symbol: M6pr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007700.1 | NP_001007701.1 | 4 - 5 | 611 | 60 | |
| XM_008763247.1 | XP_008761469.1 | 4 - 5 | 1663 | 60 | ||
| XM_008763248.1 | XP_008761470.1 | 4 - 5 | 676 | 60 | ||
| GenBank mRNA | BC079226.1 | - | 4 - 5 | 611 | 7132318 | 60 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100909548
Genomic Map