Assay Details
| Gene Symbol: Strap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011969.1 | NP_001011969.1 | 1 - 2 | 319 | 85 | |
| GenBank mRNA | BC083714.1 | - | 1 - 2 | 319 | 7190676 | 85 |
| Gene Symbol: LOC100910575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749864.3 | XP_003749912.2 | 1 - 2 | 340 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Strap
Genomic Map