Assay Details
| Gene Symbol: Zfp248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729406.4 | XP_002729452.2 | 2 - 3 | 1261 | 109 | |
| XM_006237213.2 | XP_006237275.1 | 3 - 4 | 1304 | 109 | ||
| GenBank mRNA | AY724530.1 | - | 516 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002729406.4 transcripts.
Set Membership: |
|

Zfp248
Genomic Map