Assay Details
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009486.1 | NP_001009486.1 | 5 - 6 | 590 | 87 | |
| XM_008763404.1 | XP_008761626.1 | 4 - 5 | 567 | 87 | ||
| GenBank mRNA | AY649835.1 | - | 5 - 6 | 590 | 87 | |
| BC085917.1 | - | 553 | 7108565 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klra1
Genomic Map