Assay Details
| Gene Symbol: Klrb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010964.1 | NP_001010964.1 | 2 - 3 | 204 | 75 | |
| XM_006237414.2 | XP_006237476.1 | 2 - 3 | 264 | 75 | ||
| GenBank mRNA | EF100677.1 | - | 2 - 3 | 188 | 75 | |
| EF100681.1 | - | 2 - 3 | 188 | 75 | ||
| EF100683.1 | - | 2 - 3 | 75 | |||
| FJ416340.1 | - | 2 - 3 | 209 | 75 | ||
| FQ232553.1 | - | 1 - 2 | 67 | 75 | ||
| M62891.1 | - | 2 - 3 | 204 | 75 | ||
| Gene Symbol: RGD2301395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135687.1 | NP_001129159.1 | 2 - 3 | 188 | 75 | |
| GenBank mRNA | FJ158033.1 | - | 2 - 3 | 188 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klrb1a
Genomic Map