Assay Details
| Gene Symbol: LOC502894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025064.1 | NP_001020235.1 | 2 - 3 | 237 | 63 | |
| XM_006237501.2 | XP_006237563.1 | 3 - 4 | 316 | 63 | ||
| GenBank mRNA | BC093610.1 | - | 2 - 3 | 237 | 7319640 | 63 |
| FQ217296.1 | - | 3 - 4 | 278 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC093610.1,NM_001025064.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC502894
Genomic Map