Assay Details
| Gene Symbol: Terf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012464.1 | NP_001012482.1 | 1 - 2 | 278 | 72 | |
| XM_006237729.2 | XP_006237791.1 | 1 - 2 | 338 | 72 | ||
| GenBank mRNA | BC089888.1 | - | 1 - 2 | 278 | 7374057 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Terf1
Genomic Map