Assay Details
| Gene Symbol: Cpsf3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033892.1 | NP_001029064.1 | 9 - 10 | 1044 | 63 | |
| XM_006239549.2 | XP_006239611.1 | 9 - 10 | 1086 | 63 | ||
| XM_006239550.2 | XP_006239612.1 | 9 - 10 | 1086 | 63 | ||
| XM_006239551.2 | XP_006239613.1 | 9 - 10 | 1086 | 63 | ||
| XM_006239552.2 | XP_006239614.1 | 9 - 10 | 1114 | 63 | ||
| XM_006239553.2 | XP_006239615.1 | 7 - 8 | 943 | 63 | ||
| GenBank mRNA | BC105303.1 | - | 9 - 10 | 1044 | 7110348 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Cpsf3l
Genomic Map