Assay Details
| Gene Symbol: Lin28a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109269.1 | NP_001102739.1 | 2 - 3 | 306 | 62 | |
| XM_008764170.1 | XP_008762392.1 | 2 - 3 | 321 | 62 | ||
| Gene Symbol: LOC503116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240815.2 | XP_006240877.1 | 2 - 3 | 244 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lin28a
Genomic Map