Assay Details
| Gene Symbol: Lin28a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109269.1 | NP_001102739.1 | 3 - 4 | 485 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
germ cell development
biological_process
miRNA metabolic process
miRNA catabolic process
stem cell population maintenance
pre-miRNA processing
RNA 3'-end processing
positive regulation of neuron differentiation
negative regulation of glial cell differentiation
positive regulation of translation
stem cell differentiation
regulation of gene silencing by miRNA
positive regulation of cell proliferation involved in kidney development
molecular_function
DNA binding
RNA binding
mRNA binding
zinc ion binding
translation initiation factor binding
miRNA binding
regulation of transcription, DNA-templated
germ cell development
biological_process
miRNA metabolic process
miRNA catabolic process
stem cell population maintenance
pre-miRNA processing
RNA 3'-end processing
positive regulation of neuron differentiation
negative regulation of glial cell differentiation
positive regulation of translation
stem cell differentiation
regulation of gene silencing by miRNA
positive regulation of cell proliferation involved in kidney development
molecular_function
DNA binding
RNA binding
mRNA binding
zinc ion binding
translation initiation factor binding
miRNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P22058 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P368901 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P793551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41392 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P368901 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P22058 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P793551 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P368901 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P793551 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P22058 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P793551 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P22058 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P368901 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P227483 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P368901 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8315 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49117 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10880427 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880428 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10862856 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880426 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10862855 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10880425 | - | - |
| Rat Genome 230 2.0 Array | 1393904_at | - | - |
| Rat Genome 230 2.0 Array | 1393904_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500028461.rn.1 | TC0500003798.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500028462.rn.1 | TC0500003798.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500028459.rn.1 | TC0500003798.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500028460.rn.1 | TC0500003798.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393904_at | - | - |
| Rat Expression Set 230 | 1393904_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10880425 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21797306 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650083 | - | - |

Lin28a
Genomic Map