Assay Details
| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_342941.7 | XP_342942.5 | 4 - 5 | 520 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Catsper4| Gene Symbol: Catsper4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_342941.7 | XP_342942.5 | 4 - 5 | 520 | 64 | |
Set Membership: |
|