Assay Details
| Gene Symbol: Akirin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030054.1 | NP_001025225.1 | 2 - 3 | 457 | 63 | |
| GenBank mRNA | CV107792.1 | - | 2 - 3 | 457 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akirin1
Genomic Map