Assay Details
| Gene Symbol: RGD1562036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_232777.6 | XP_232777.4 | 3 - 4 | 488 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562036| Gene Symbol: RGD1562036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_232777.6 | XP_232777.4 | 3 - 4 | 488 | 97 | |
Set Membership: |
|