Assay Details
| Gene Symbol: Zbtb8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107913.1 | NP_001101383.1 | 3 - 4 | 1154 | 108 | |
| XM_006238931.2 | XP_006238993.1 | 3 - 4 | 1570 | 108 | ||
| GenBank mRNA | BC161892.1 | - | 3 - 4 | 1087 | 7191093 | 108 |
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb8a
Genomic Map