Assay Details
| Gene Symbol: Acot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130756.3 | NP_570112.3 | 3 - 4 | 886 | 79 | |
| GenBank mRNA | AF452100.1 | - | 3 - 4 | 536 | 79 | |
| BC078751.1 | - | 3 - 4 | 880 | 7114090 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acot8
Genomic Map